missing translation for 'onlineSavingsMsg'
Learn More

Tropylium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. T11211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11211G 1 g
T11215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11211G Supplier TCI America Supplier No. T11211G

Chemical Identifiers

CAS 27081-10-3
Molecular Formula C7H7BF4
Molecular Weight (g/mol) 177.94
MDL Number MFCD00013402
InChI Key SQVQHTIKOZVGLR-UHFFFAOYSA-N
Synonym tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t
PubChem CID 152926
IUPAC Name cyclohepta-2,4,6-trien-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.[CH+]1C=CC=CC=C1

Specifications

Quantity 1 g
UN Number 1759
Formula Weight 177.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tropylium Tetrafluoroborate
TSCA Yes
Recommended Storage Refrigerator