missing translation for 'onlineSavingsMsg'
Learn More

Tropylium tetrafluoroborate, 97%

Catalog No. AAA1404514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404514 25 g
AAA1404506 5 g
AAA1404522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404514 Supplier Thermo Scientific Chemicals Supplier No. A1404514

CAS: 27081-10-3 | C7H7BF4 | 177.94 g/mol

Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant or reagent involved in Retro-Buchner reactions for synthesis of arylcyclopropanes, α-Cyanation of imines, electrochemical redox transformations, oxidative C-H activation of aliphatic aldehydes.

Solubility
Soluble in hot methanol.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, strong bases, water/ moisture.

Chemical Identifiers

CAS 27081-10-3
Molecular Formula C7H7BF4
Molecular Weight (g/mol) 177.94
MDL Number MFCD00013402
InChI Key SQVQHTIKOZVGLR-UHFFFAOYSA-N
Synonym tropylium tetrafluoroborate, cycloheptatrienyl tetrafluoroborate, cyclohepta-1,3,5-triene tetrafluoroborate, cycloheptatrienylium, tetrafluoroborate 1-, tropylium tetrafluoroborate 1-, cycloheptatrienylium, tetrafluoroborate 1-1:1, cyclohepta-2,4,6-trienylium tetrafluoroborat, tropylium tetrafluoroborate, purum t
PubChem CID 152926
IUPAC Name cyclohepta-1,3,5-triene;tetrafluoroborate
SMILES F[B-](F)(F)F.[CH+]1C=CC=CC=C1

Specifications

Melting Point ∼240°C (decomposition)
Quantity 25 g
UN Number UN1759
Beilstein 3574512
Sensitivity Moisture sensitive
Solubility Information Soluble in hot methanol.
Formula Weight 177.94
Percent Purity 97%
Chemical Name or Material Tropylium tetrafluoroborate
Hazard Category H314-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 248-214-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only