missing translation for 'onlineSavingsMsg'
Learn More

Tropone, 97%

Catalog No. AAL0047303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0047303 1 g
AAL0047306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0047303 Supplier Thermo Scientific Chemicals Supplier No. L0047303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 539-80-0 | C7H6O | 106.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 539-80-0
Molecular Formula C7H6O
Molecular Weight (g/mol) 106.124
MDL Number MFCD00014331
InChI Key QVWDCTQRORVHHT-UHFFFAOYSA-N
Synonym tropone, 2,4,6-cycloheptatrien-1-one, tropon, unii-co48x7sufh, co48x7sufh, cycloheptatrienone, 1-cyclohepta-2,4,6-trienone, 2,4,6-cycloheptatriene-1-one
PubChem CID 10881
IUPAC Name cyclohepta-2,4,6-trien-1-one
SMILES C1=CC=CC(=O)C=C1

Specifications

Density 1.094
Boiling Point 91°C to 92°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 1 g
Beilstein 1902335
Formula Weight 106.12
Percent Purity 97%
Chemical Name or Material Tropone
EINECSNumber 208-725-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only