missing translation for 'onlineSavingsMsg'
Learn More

Tropolone 98.0+%, TCI America™

Catalog No. p-7136359
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
T06061G 1 g
T06065G 5 g
T060625G 25 g
3 options

Catalog No. T06061G

Supplier: TCI America T06061G

Only null left

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00004158
InChI Key MDYOLVRUBBJPFM-UHFFFAOYSA-N
Synonym tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
PubChem CID 10789
ChEBI CHEBI:79966
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES C1=CC=C(C(=O)C=C1)O

Specifications

Melting Point 53°C
Quantity 1 g
Formula Weight 122.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tropolone
EINECSNumber (9)-0885
RTECSNumber GU4075000
TSCA No
Recommended Storage Refrigerator