missing translation for 'onlineSavingsMsg'
Learn More

Tropolone, 98%

CAS: 533-75-5 | C7H6O2 | 122.12 g/mol

$303.86 - $303.86

Chemical Identifiers

CAS 533-75-5
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004158
InChI Key MDYOLVRUBBJPFM-UHFFFAOYSA-N
Synonym tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
PubChem CID 10789
ChEBI CHEBI:79966
IUPAC Name 2-hydroxycyclohepta-2,4,6-trien-1-one
SMILES C1=CC=C(C(=O)C=C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140580050
SDS View Documents
Thermo Scientific Chemicals
140580050
5 g Glass bottle
Each for $303.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

533-75-5
122.12
MDYOLVRUBBJPFM-UHFFFAOYSA-N
10789
2-hydroxycyclohepta-2,4,6-trien-1-one
C7H6O2
MFCD00004158
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
CHEBI:79966
C1=CC=C(C(=O)C=C1)O

Specifications

533-75-5
Beige-Cream to White
>110°C
97.5% min. (GC)
C7H6O2
5 g
tropolone, purpurocatechol, 2-hydroxycyclohepta-2,4,6-trienone, 2,4,6-cycloheptatrien-1-one, 2-hydroxy, 2-hydroxytropone, 2-hydroxy-2,4,6-cycloheptatrien-1-one, 2-hydroxy-2,4,6-cycloheptatrienone, unii-7l6dl16p1t, ccris 6609, tropomyosins
MDYOLVRUBBJPFM-UHFFFAOYSA-N
2-hydroxycyclohepta-2,4,6-trien-1-one
10789
122.12
Crystalline Powder
50.0°C to 54.0°C
115.0°C (12.0 mmHg)
Authentic
Glass bottle
MFCD00004158
15, 9963
Solubility in water: soluble in water. Other solubilities: soluble in ethyl ether, acetone, organic solvents
C1=CC=C(C(=O)C=C1)O
122.12
CHEBI:79966
98%
Tropolone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-577-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only