missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tropicamide, 99+%
Description
A muscarinic antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 g |
| Description | White |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | A muscarinic antagonist |
| Formula Weight | 284.35g/mol |
| Melting Point | 98°C |
| Product Type | Signal Transduction Reagent |
| Form | Powder |
| Assay | ≥99% |
RUO – Research Use Only