missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tropaeolin O, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Visual Transition Interval | from pH 11.1 (yellow) to pH 12.7 (orange) |
| Beilstein | 16, 275 |
| Merck Index | 15, 9954 |
| Color | Brown to Rust |
| Solubility Information | Solubility in water: Soluble. Other solubilities: Soluble in alcohol |
| Quantity | 100 g |
| Physical Form | Powder |
| Formula Weight | 316.26 |
| Show More |
Safety and Handling
| EINECSNumber | 208-924-8 |
| RTECSNumber | DB6145180 |
| TSCA | TSCA |
RUO – Research Use Only