missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tropaeolin O

Catalog No. AA3870804
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3870804 2 g
AA3870809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3870804 Supplier Thermo Scientific Chemicals Supplier No. 03870804

CAS: 547-57-9 | C12H10N2NaO5S | 317.271 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym Acid Orange 6; C.I. 14270
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]

Specifications

Merck Index 14,9775
Solubility Information Soluble in water and alcohol
Formula Weight 316.27
Color Brown to Orange
Quantity 2 g
Physical Form Powder
Chemical Name or Material Tropaeolin O
EINECSNumber 208-924-8
RTECSNumber DB6145180
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only