Learn More
Enzo Life Sciences TROP2 (human) monoclonal antibody (01) (100 l)
Supplier: Enzo ENZABS3800100
Clone: 01. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human TROP2 extracellular domain. UniProt: P09758. Species Reactivity: Human. Crossreactivity: Does not cross-react with mouse or rat TROP2. Applications: ELISA. Flow Cytometry, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. 0.2 M filtered solution in PBS. Handling: Sodium azide is recommended to avoid contamination (final. Concentration 0.05%-0.1%) It is toxic to cells and should be disposed of properly. Avoid freeze-thaw cycles.