missing translation for 'onlineSavingsMsg'
Learn More

Trolamine, NF, 99-107.4%, Spectrum™ Chemical

Catalog No. p-7158424
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607603 500 mL Amber Glass Bottle
18607604 6 x 500 mL Amber Glass Bottle
18607605 4 L Amber Glass Bottle
18607606 4 x 4 L Amber Glass Bottle
18607607 20 L Poly Drum
18607608 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18607603 Supplier Spectrum Chemical Mfg Cor Supplier No. TR143500MLGL
Only null left

TR143, 102-71-6, C6H15NO3

Spectrum™ Chemical Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

CAS Max % 1
Ignition Residue 0.0005
Packaging Amber Glass Bottle
Refractive Index 1.481-1.486
Quantity 500 mL
Percent Purity 99 to 107.4%
Grade NF