Learn More
Troemner™ Talboys™ Ultrajaws Heavy-Duty Clamps
Enables secure gripping and positioning with added strength and durability
Supplier:  Troemner™ 30392250
 View more versions of this product
Catalog No. 02-216-240
Only null left
Explore available promotions
Description
Description
Enables secure gripping and positioning with added strength and durability
  • Three-pronged grip with large grip adjustment range
  • Closed yoke construction that minimizes contamination and corrosion of internal components
  • Precise pressure regulation when gripping glassware surfaces reduces the chance of breakage
  • Constructed of nickel-plated zinc with vinyl sleeves
  • For temperatures above 100°C, fiberglass covers
An additional holder must be purchased in order to attach clamps to frames or other apparatus.
Specifications
Specifications
Heavy Duty Clamp
Use in holding and securing Glassware with precision ground Glass joints
12.7 cm
7.18 cm
Nickel, Zinc
5 in.
Large
2.83 in.
Product Suggestions
Product Suggestions