Learn More
Troemner™ Talboys™ Tube Hosecocks
Flow control devices deliver accurate regulation or interruption of fluid flow.
$159.73 - $159.73
Specifications
| Material | Nickel-plated Zinc |
|---|---|
| Includes | Adjustment screw with oversized head |
| For Use With (Application) | Accurate regulation |
| Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (Metric) | Price | Quantity | |||||
|
02-300-207
|
Troemner™
30392210 |
62 mm |
Each for $159.73
|
|
|||||
Description
One-hand operation
- Convex bearing surfaces and rounded edges protect tubing
- Adjustment screw with oversized head for accurate regulation
- Built-in side lugs for foot mounting
- Corrossion and rust resistant
- Nickel-plated zinc construction
Specifications
| Nickel-plated Zinc | |
| Adjustment screw with oversized head | |
| Accurate regulation |