Learn More
Troemner™ Talboys™ Labjaws™ Chain Clamp
Holds large round or irregular shaped objects firmly, yet gently to lab-frames and rods
Description
- Quick and secure slip-on chain connection with large, easy-to-turn adjusting knob
- Extension arm allows user to vary distance from the frame
- Available as stainless steel clamp, constructed entirely of stainless steel with electro-polished finish or nickel-plated zinc clamp with strong, chromed-brass chain
Chemical Identifiers
| Chain Clamp | |
| 5 in. | |
| 12.7 cm | |
| Large |
| Holds round or irregular shaped objects to lab-frames and rods | |
| 5 in. | |
| 12.7 cm |
Specifications
| Chain Clamp | |
| Quick and secure slip-on chain connection; Simple-turn adjusting knob | |
| 5 in. | |
| 12.7 cm | |
| Large | |
| 11 in. |
| Nickel, Zinc, Chromed Brass | |
| Holds round or irregular shaped objects to lab-frames and rods | |
| 5 in. | |
| 12.7 cm | |
| 27.9 cm |