Learn More
Troemner™ 17 in. Support Rod Kit
Catalog No. 02214208
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
02214208 Stainless Steel
1 options
Catalog No. 02214208 Supplier Troemner™ Supplier No. 30400050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Creates an adjustable support stand on the upper deck of a 6 x 6 in. , 8 x 8 in. , 10 x 10 in. , 12 x 12 in. , or 16 x 16 in. lab-lift
  • Ideal for mounting thermometer clamps, temperature probes, as well as flask and column clamps
Specifications
Material Stainless Steel
Includes 17 in. stainless steel rod, 1 jam nut
Product Type Support Rod Kit
For Use With (Application) For mounting various items such as thermometer clamps, temperature probes, flask and column clamps