Learn More
Troemner™ Regular Hosecock, Stainless Steel
Description
- Flow control device resists corrosion and rust
- Offer easy one-hand operation
- Convex bearing surfaces and rounded edges protect tubing
- Operated with a simple squeeze
- Nickel-plated construction
- Clamp dimensions: W x H -open 37 x 62mm (1.46 x 2.44 in.)
Specifications
Specifications
| Type | Regular Hosecock |
| Material | Stainless Steel |