Learn More
SignalChem Biotech TPR-TRKA (TRK-T1), Active 50ug
Catalog No. 502574019
Encompass
Recombinant human fusion protein TPR (1-191)-TRKA (399-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502574019 Supplier SignalChem Biotech Supplier No. N1619BG50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human fusion protein TPR (1-191)-TRKA (399-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.