SignalChem Biotech TFG-MET (Tex5Mex15), Active 500ug
Catalog No. 502574134
Recombinant human TFG (1-193a.a., exons1-5)-MET (1010a.a.-end, exons15-21) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human TFG (1-193a.a., exons1-5)-MET (1010a.a.-end, exons15-21) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.