missing translation for 'onlineSavingsMsg'
Learn More

Trizma™ Hydrochloride Buffer Solution, BioUltra, For Molecular Biology, pH 7.4, MilliporeSigma™ Supelco™

Catalog No. 111002698
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002698 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002698 Supplier Merck Emd Millipore Supplier No. 933131L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer component (SET buffer) in the preparation of RNA

Specifications

Density 1.05 g/mL (at 20°C)
MDL Number MFCD00012590
Quantity 1 L
Synonym Tris(hydroxymethyl)aminomethane hydrochloride buffer solution
Grade For molecular biology
Physical Form Liquid
Product Line Trizma

Trizma is a registered trademark of Sigma-Aldrich Co. LLC