missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Ricca Chemical

Catalog No. p-3339879
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8698516 500 mL Natural Poly Bottle
R86985001A 1 L Natural Poly Bottle
R86985004A 4 L Natural Poly Bottle
869855 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 8698516 Supplier Ricca Chemical Company Supplier No. 8698.516

Wetting Agent

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless
Physical Form Viscous Liquid
CAS Min % 0.98
Boiling Point 270°C
CAS Max % 1.0
Melting Point 6°C
Concentration 100%
pH 6
Quantity 500 mL
Packaging Natural Poly Bottle
Grade Laboratory
Chemical Name or Material Triton™ X-100 Wetting Agent
Show More Show Less
ShelfLife 24 months

Triton is a trademark of Union Carbide Corporation, a wholly owned subsidiary of The Dow Chemical Company.