missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100

Catalog No. AAA16046AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16046AP 500 mL
AAA16046AE 100 mL
AAA160460F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA16046AP Supplier Thermo Scientific Chemicals Supplier No. A16046AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Boiling Point >250°C
Melting Point ∽5°C
Density 1.070
Quantity 500 mL
Flash Point 180°C (356°F)
Refractive Index 1.49
Solubility Information Miscible with water.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
Formula Weight ca. 625
Chemical Name or Material Triton X-100
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
RTECSNumber MD0907600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only