missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton X-100 Surfactant, MilliporeSigma™
Supplier: Technidata TX15683
| Quantity | 4 L |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| C16H26O2 | |
| MFCD00132505 | |
| Octylphenol Decaethylene Glycol Ether | |
| 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol |
Specifications
| Triton X-100 | |
| Surfactant | |
| Liquid | |
| Glass Bottle | |
| C34H62O11 | |
| Octylphenol Decaethylene Glycol Ether | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
| 250.38 | |
| 646.86 |
| Undesignated | |
| 9002-93-1 | |
| 4 L | |
| C16H26O2 | |
| MFCD00132505 | |
| JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
| 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol | |
| 5590 |
Safety and Handling
Recommended Storage : Room Temperature