missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triton X-100 Surfactant, MilliporeSigma™
Specifications
Specifications
| Color | Undesignated |
| Physical Form | Liquid |
| pH | 5 to 8 |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Linear Formula | C34H62O11 |
| Formula Weight | 646.86 |
| Chemical Name or Material | Triton X-100, Surfactant |
Safety and Handling
| Recommended Storage | Room Temperature |