missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100™ Solution, 2% v/v, Reagents

Catalog No. 01333447
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333447 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333447 Supplier Labchem Inc Supplier No. CT104200500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triton X-100™ Solution, 2% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Triton X-100™ Solution, 2% v/v
Color Colorless
CAS 7732-18-5, 9002-93-1
Physical Form Liquid
Boiling Point 100°C
Quantity 500 mL
Packaging Poly Bottle
Specific Gravity 1.0013
Synonym Octoxynol, Octoxinol
Percent Purity 2%
Grade Reagent
Show More Show Less