missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100, Molecular Biology Reagent Grade, MP Biomedicals™

Catalog No. ICN19485450
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19485450 50 mL
ICN19485480 100 mL
ICN19485483 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19485450 Supplier MP Biomedicals Inc Supplier No. 0219485450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, 4-(1, 1, 3
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless to Yellow
Physical Form Viscous Liquid
Density 8.9 lb./gal.
Quantity 50 mL
Grade Molecular Biology
Chemical Name or Material Triton™ X-100
Recommended Storage Store at room temperature (15° to 30°C)