missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-100 (Electrophoresis), Fisher BioReagents™

Catalog No. BP151100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP151100 100 mL Glass Bottle
BP151500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP151100 Supplier Fisher BioReagents Supplier No. BP151 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For solubilization and permeabilization of membranes

This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyethylene Glycol p-tert-Octylphenyl Ether
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Liquid
Boiling Point 270°C
Melting Point 6°C
Cloud Point 63 to 69°C
Density 1.067g/cm³
pH 6 to 8
Quantity 100 mL
Identification Pass Test
Packaging Glass Bottle
Linear Formula 4-(C8H17) C6H4 (OCH2CH2)n OH (n∽10)
Merck Index 15, 6850
Solubility Information Soluble in water
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Formula Weight 646.86
Grade Electrophoresis
Chemical Name or Material Triton™ X-100
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artificial respiration with the aid of a pocket mask equipped with a one way valve or other proper respiratory medical device. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:1
ShelfLife 2 years
Recommended Storage RT