accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Facility Safety and Maintenance
Cleaning Supplies and Equipment
Cleaners and Disinfectants
Laboratory Surfactants and Wetting Agents
MilliporeSigma™ Triton X-100 Detergent, Calbiochem™,
Structure Search
Print
MilliporeSigma™ Triton X-100 Detergent, Calbiochem™,
Catalog No.
p-7082991
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Fiber Carton
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
6484621KG
1 kg
Glass Bottle
6484623KG
3 kg
Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No.
6484621KG
Supplier
MilliporeSigma™
Supplier No.
6484621KG
Sign In
or
Register
to check your price and availability.
Quantity
Only null left
Add to cart
Add to cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
9002-93-1
Molecular Formula
C16H26O2
Molecular Weight (g/mol)
250.38
MDL Number
MFCD00132505
InChI Key
JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym
Polyethylene Glycol-p-isooctylphenyl Ether
PubChem CID
5590
IUPAC Name
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Specifications
Color
Colorless to Yellow
Quantity
1 kg
Packaging
Glass Bottle
Product Suggestions
Documents