missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Triton™X-100 Surfactant, OmniPur™, Calbiochem™,

Catalog No. p-7082900
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
9400100ML 100 mL Glass Bottle
94101L 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9400100ML Supplier MilliporeSigma™ Supplier No. 9400100ML
Only null left

Used for solubilization of membrane proteins, enzymology, analytical and diagnostic work, as well as chromatography and electrophoresis

Recommended Storage

Ok to freeze

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Viscous liquid
pH 6.0 to 8.0
Quantity 100 mL
Packaging Glass Bottle
Water <0.5%
Grade Molecular Biology
storageNote1 Ok to freeze