missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Triton™X-100 Surfactant, OmniPur™, Calbiochem™,

Catalog No. 9400100ML
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
9400100ML 100 mL Glass Bottle
94101L 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9400100ML Supplier MilliporeSigma™ Supplier No. 9400100ML

Used for solubilization of membrane proteins, enzymology, analytical and diagnostic work, as well as chromatography and electrophoresis

Recommended Storage

Ok to freeze

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Viscous liquid
pH 6.0 to 8.0
Quantity 100 mL
Packaging Glass Bottle
Water <0.5%
Grade Molecular Biology
storageNote1 Ok to freeze