missing translation for 'onlineSavingsMsg'
Learn More

Tritolyl phosphate, mixture of isomers

Catalog No. p-7023648
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17433AP 500 mL
AAA174330F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA17433AP Supplier Thermo Scientific Chemicals Supplier No. A17433AP
Only null left

CAS: 1330-78-5 | C21H21O4P

Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tritolyl phosphate, mixture of isomers is used as plasticizer in nitrocellulose, acrylate lacquers, and in polyvinyl chloride. It is a fire retardant for plastics and rubbers. air filter medium, solvent mixtures and heat exchange medium. It is employed as a gasoline additive and as a lead scavenger for tetraethyllead. Further, it acts as a solvent for extraction, especially for nitrocellulose and polymers. In addition to this, it is used for the waterproofing of materials, an antiwear and extreme pressure additive in lubricants and hydraulic fluids.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, sulfuric acid, caustics, ammonia, aliphatic amines, alkanolamines, isocyanates, alkylene oxides, epichlorohydrin and nitromethane.

Chemical Identifiers

CAS 1330-78-5
Molecular Formula C21H21O4P
MDL Number MFCD02102116
Synonym Tricresyl phosphate; TCP

Specifications

Density 1.167
Boiling Point 265°C (10 mmHg)
Flash Point 250°C (482°F)
Odor Odorless
Linear Formula (CH3C6H4O)3P=O
Quantity 500 mL
UN Number UN2574
Merck Index 14,9763
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 368.37
Percent Purity 98%
Chemical Name or Material Tritolyl phosphate, mixture of isomers, ortho-isomers <1%
Show More Show Less
Hazard Category H361fd
Hazard Statement GHS H Statement
H370
Causes damage to organs.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TRICRESYL PHOSPHATE
EINECSNumber 215-548-8
RTECSNumber TD0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only