missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trisodium Nitrilotriacetate Monohydrate 98.0+%, TCI America™
$35.41 - $82.02
Chemical Identifiers
| CAS | 18662-53-8 |
|---|---|
| Molecular Formula | C6H8NNa3O7 |
| Molecular Weight (g/mol) | 275.099 |
| MDL Number | MFCD00150789 |
| InChI Key | HERBOKBJKVUALN-UHFFFAOYSA-K |
| Synonym | trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate |
| PubChem CID | 29194 |
| IUPAC Name | trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate |
| SMILES | C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] |
Chemical Identifiers
| 18662-53-8 | |
| 275.099 | |
| HERBOKBJKVUALN-UHFFFAOYSA-K | |
| 29194 | |
| C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] |
| C6H8NNa3O7 | |
| MFCD00150789 | |
| trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate | |
| trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate |
Specifications
| 18662-53-8 | |
| 10 to 12 (50g/L aq. 25°C) | |
| MFCD00150789 | |
| trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate | |
| C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+] | |
| 275.099 | |
| 257.08 | |
| Crystalline Powder |
| White | |
| C6H8NNa3O7 | |
| 25 g | |
| HERBOKBJKVUALN-UHFFFAOYSA-K | |
| trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate | |
| 29194 | |
| ≥98.0% (T) | |
| Trisodium Nitrilotriacetate Monohydrate |
Safety and Handling
EINECSNumber : (2)-1277
RTECSNumber : AJ1070000
TSCA : Yes