missing translation for 'onlineSavingsMsg'
Learn More

Trisodium Nitrilotriacetate Monohydrate 98.0+%, TCI America™

Catalog No. N010125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010125G 25 g
N0101500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N010125G Supplier TCI America Supplier No. N010125G

Chemical Identifiers

CAS 18662-53-8
Molecular Formula C6H8NNa3O7
Molecular Weight (g/mol) 275.099
MDL Number MFCD00150789
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Synonym trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Color White
pH 10 to 12 (50g/L aq. 25°C)
Quantity 25 g
Formula Weight 257.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trisodium Nitrilotriacetate Monohydrate
EINECSNumber (2)-1277
RTECSNumber AJ1070000
TSCA Yes