missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate 98.0+%, TCI America™
$29.83 - $115.57
Chemical Identifiers
| CAS | 85715-60-2 |
|---|---|
| Molecular Formula | C10H13N2Na3O8 |
| Molecular Weight (g/mol) | 358.19 |
| MDL Number | MFCD00149675 |
| InChI Key | QZKRHPLGUJDVAR-UHFFFAOYSA-K |
| Synonym | trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate |
| PubChem CID | 16211179 |
| IUPAC Name | trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate |
| SMILES | [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
Chemical Identifiers
| 85715-60-2 | |
| 358.19 | |
| QZKRHPLGUJDVAR-UHFFFAOYSA-K | |
| 16211179 | |
| [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O |
| C10H13N2Na3O8 | |
| MFCD00149675 | |
| trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate | |
| trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate |
Specifications
| 85715-60-2 | |
| C10H13N2Na3O8 | |
| 25 g | |
| QZKRHPLGUJDVAR-UHFFFAOYSA-K | |
| trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate | |
| 16211179 | |
| ≥98.0% (T) | |
| Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate |
| White | |
| MFCD00149675 | |
| trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate | |
| [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O | |
| 358.19 | |
| 358.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1265
RTECSNumber : AH5250000
TSCA : No