missing translation for 'onlineSavingsMsg'
Learn More

Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate 98.0+%, TCI America™

Catalog No. E043025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E043025G 25 g
E0430500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E043025G Supplier TCI America Supplier No. E043025G
Only null left

Chemical Identifiers

CAS 85715-60-2
Molecular Formula C10H13N2Na3O8
Molecular Weight (g/mol) 358.19
MDL Number MFCD00149675
InChI Key QZKRHPLGUJDVAR-UHFFFAOYSA-K
Synonym trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
PubChem CID 16211179
IUPAC Name trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate
SMILES [Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 358.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate
EINECSNumber (2)-1265
RTECSNumber AH5250000
TSCA No