missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyloxy)silane 98.0+%, TCI America™

Catalog No. p-7137782
Change view
Click to view available options
Quantity:
5 mL
25 mL
Catalog No. Quantity
T35205ML 5 mL
T352025ML 25 mL
2 options

Catalog No. T35205ML

Supplier: TCI America T35205ML

Only null left

Chemical Identifiers

CAS 1873-89-8
Molecular Formula C9H27O3Si4
Molecular Weight (g/mol) 295.65
MDL Number MFCD00054865
InChI Key XAASNKQYFKTYTR-UHFFFAOYSA-N
Synonym tris trimethylsiloxy silane, tris trimethylsilyloxy silane, tris trimethylsilyloxy silicon, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, 1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxane, tris trimethylsilyloxy silyl radical, trisiloxane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy
PubChem CID 6365044
IUPAC Name 2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 185°C
Quantity 5 mL
UN Number 1993
Formula Weight 296.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyloxy)silane
EINECSNumber (7)-0476&(7)-0477