missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyloxy)ethylene 95.0+%, TCI America™

Catalog No. T16395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16395G 5 g
T163925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16395G Supplier TCI America Supplier No. T16395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69097-20-7
Molecular Formula C11H28O3Si3
Molecular Weight (g/mol) 292.60
MDL Number MFCD00011641
InChI Key FCZGHPGTZRTDNN-UHFFFAOYSA-N
Synonym tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene
PubChem CID 553067
IUPAC Name 2,2,7,7-tetramethyl-4-[(trimethylsilyl)oxy]-3,6-dioxa-2,7-disilaoct-4-ene
SMILES C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 292.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyloxy)ethylene