missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane 90.0+%, TCI America™

Catalog No. p-7136790
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14635ML 5 mL
T146325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14635ML Supplier TCI America Supplier No. T14635ML
Only null left

Chemical Identifiers

CAS 1873-77-4
Molecular Formula C9H28Si4
Molecular Weight (g/mol) 248.66
MDL Number MFCD00077893
InChI Key SQMFULTZZQBFBM-UHFFFAOYSA-N
Synonym tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
IUPAC Name 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

Specifications

Color Yellow
Physical Form Liquid
Quantity 5 mL
UN Number 1993
Formula Weight 248.66
Percent Purity ≥90.0% (GC)
Chemical Name or Material Tris(trimethylsilyl)silane [Reducing Reagent]