Learn More
Tris(trimethylsilyl)silane, 97%
Description
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.
Solubility
Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 82°C to 84°C (12mmHg) |
| Quantity | 5 g |
| Flash Point | 55°C (131°F) |
| Refractive Index | 1.49 |
| Sensitivity | Air Sensitive |
| Density | 0.806 |
| UN Number | UN1993 |
| Beilstein | 1923953 |
| Merck Index | 14,9757 |
| Solubility Information | Miscible with pentane,ether,toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only