missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane, 96%

CAS: 1873-77-4 | C9H28Si4 | 248.66 g/mol

$127.46 - $127.46

Chemical Identifiers

CAS 1873-77-4
Molecular Formula C9H28Si4
Molecular Weight (g/mol) 248.66
MDL Number MFCD00077893
InChI Key SQMFULTZZQBFBM-UHFFFAOYSA-N
Synonym tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
IUPAC Name 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291060025
SDS View Documents
Thermo Scientific Chemicals
291060025
2.5 mL Glass bottle
Each for $127.46
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

1873-77-4
248.66
SQMFULTZZQBFBM-UHFFFAOYSA-N
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
C9H28Si4
MFCD00077893
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

Specifications

82°C to 84°C (12.0mmHg)
Colorless to Yellow
2.5 mL
C9H28Si4
MFCD00077893
55°C
tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
SQMFULTZZQBFBM-UHFFFAOYSA-N
1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
248.66
Authentic
1.4890 to 1.491
0.8000g/mL
1873-77-4
Liquid
95% min. (GC)
((CH3)3Si)3SiH
15,358; 17,395
15, 9934
Solubility in water: insoluble
C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C
248.66
96%
Glass bottle
0.8
Tris(trimethylsilyl)silane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only