missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl)silane, 96%

Catalog No. AC291060025
Change view
Click to view available options
Quantity:
2.5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291060025 2.5 mL Glass bottle
AC291060250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC291060025 Supplier Thermo Scientific Chemicals Supplier No. 291060025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1873-77-4 | C9H28Si4 | 248.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 1873-77-4
Molecular Formula C9H28Si4
Molecular Weight (g/mol) 248.66
MDL Number MFCD00077893
InChI Key SQMFULTZZQBFBM-UHFFFAOYSA-N
Synonym tris trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane, tris trimethylsilyl silicon, ttmss, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl-trisilane, hypersilyl, tris trimethylsilyl silyl radical, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilane #, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl trisilan-2-yl
IUPAC Name 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
SMILES C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C

Specifications

Boiling Point 82°C to 84°C (12.0mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 2.5 mL
Flash Point 55°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4890 to 1.491
Specific Gravity 0.8
Assay Percent Range 95% min. (GC)
Density 0.8000g/mL
Linear Formula ((CH3)3Si)3SiH
Fieser 15,358; 17,395
Merck Index 15, 9934
Solubility Information Solubility in water: insoluble
Formula Weight 248.66
Percent Purity 96%
Chemical Name or Material Tris(trimethylsilyl)silane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

RUO – Research Use Only