missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) Phosphite 95.0+%, TCI America™

Catalog No. p-7135013
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12175ML 5 mL
P121725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12175ML Supplier TCI America Supplier No. P12175ML
Only null left

Chemical Identifiers

CAS 1795-31-9
Molecular Formula C9H27O3PSi3
Molecular Weight (g/mol) 298.541
MDL Number MFCD00015588
InChI Key VMZOBROUFBEGAR-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
PubChem CID 137213
IUPAC Name tris(trimethylsilyl) phosphite
SMILES C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 73°C
Quantity 5 mL
Formula Weight 298.54
Percent Purity ≥95.0% (NMR)
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) Phosphite