missing translation for 'onlineSavingsMsg'
Learn More

Tris(trimethylsilyl) phosphite, 92%

Catalog No. AC428521000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428521000 100 g Glass bottle
AC428520050 5 g Glass bottle
AC428520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC428521000 Supplier Thermo Scientific Chemicals Supplier No. 428521000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1795-31-9 | C9H27O3PSi3 | 298.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1795-31-9
Molecular Formula C9H27O3PSi3
Molecular Weight (g/mol) 298.54
MDL Number MFCD00015588
InChI Key VMZOBROUFBEGAR-UHFFFAOYSA-N
Synonym tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
PubChem CID 137213
IUPAC Name tris(trimethylsilyl) phosphite
SMILES C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Density 0.8930g/mL
Boiling Point 78.0°C to 81.0°C (8.3 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4080 to 1.4110 (20°C, 589nm)
Linear Formula P[OSi(CH3)3]3
Quantity 100 g
Specific Gravity 0.893
Solubility Information Solubility in water: reacts.
Formula Weight 298.54
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) phosphite
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only