missing translation for 'onlineSavingsMsg'
Learn More

Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)

Catalog No. p-7080461
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8848106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8848106 Supplier Thermo Scientific Chemicals Supplier No. 8848106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34830-11-0 | C36H45EuF9O6 | 896.7 g/mol

Optically active NMR shift reagent. Used with Ag(fod) for the enantiomeric resolution of sec-butylisothiouronium chloride. Also reactant involved in synthesis of europium acetyl camphorate phosphine oxide complexes and reactions with 1,10-N,N-phenanthroline dioxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34830-11-0
Molecular Formula C36H45EuF9O6
Molecular Weight (g/mol) 896.7
MDL Number MFCD00074803
InChI Key YXJPWWGLCOPUGI-UFRADDTFSA-N
Synonym eu tfc 3, eu facam 3, europium tris 3-trifluoromethylhydroxymethylene---camphorate, europium tris 3-trifluoromethylhydroxymethylene-+-camphorate, tris 3-trifluoromethylhydroxymethylene-d-camphorato europium iii, tris 3-trifluoroacetyl-d-camphorato europium iii, europium iii tris 3-trifluoromethylhydroxymethylene-d-camphorate
PubChem CID 71310195
IUPAC Name europium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.[Eu]

Specifications

Melting Point 195°C to 198°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 893.68
Physical Form Crystalline
Chemical Name or Material Tris (trifluoromethylhydroxymethylene-d-camphorato)europium (III)
EINECSNumber 252-232-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only