missing translation for 'onlineSavingsMsg'
Learn More

Tris(trifluoro-2,4-pentanedionato)aluminum(III) 98.0+%, TCI America™

Catalog No. A12675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12675G 5 g
A126725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12675G Supplier TCI America Supplier No. A12675G

Chemical Identifiers

CAS 14354-59-7
Molecular Formula C15H15AlF9O6
Molecular Weight (g/mol) 489.25
MDL Number MFCD00042062
InChI Key DOYKOKXTRGISGV-DJFUMVPSSA-N
Synonym Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt

Specifications

Melting Point 121°C
Quantity 5 g
Formula Weight 486.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(trifluoro-2,4-pentanedionato)aluminum(III)