missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ TRIS, OmniPur™, Calbiochem™,
Supplier: MilliporeSigma™ 92305KG
Specifications
| Tris | |
| 168°C to 172°C | |
| 10.2 to 10.6 | |
| Odorless | |
| 5 kg | |
| Water: Soluble | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| ≥99.8% |
| 77-86-1 | |
| White | |
| 219°C to 220°C | |
| C4H11NO3 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.1 | |
| Solid |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.