missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ TRIS, OmniPur™, Calbiochem™,
Specifications
Specifications
| Melting Point | 168°C to 172°C |
| Color | White |
| pH | 10.2 to 10.6 |
| Boiling Point | 219°C to 220°C |
| Odor | Odorless |
| Quantity | 500 g |
| Solubility Information | Water: Soluble |
| Formula Weight | 121.1 |
| Percent Purity | ≥99.8% |
| Grade | Not Available |
| Show More |
Safety and Handling
| Recommended Storage | Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. |