missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(hydroxymethyl)aminomethane, USP, EP, bioCERTIFIED, 99-100.5%, Spectrum™ Chemical
Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.
$823.21 - $823.21
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.14 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | NC(CO)(CO)CO |
Description
- Full testing including bioburden, endotoxin and elemental analysis
- Comprehensive regulatory and scientific documentation
- Change control and batch traceabilitiy
- Biopharm industry standard CFR 210/211 cGMP
Chemical Identifiers
| 77-86-1 | |
| 121.14 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| NC(CO)(CO)CO |
Specifications
| 77-86-1 | |
| 10 to 11.5 | |
| 0.5% | |
| Plastic Bottle | |
| 1 kg | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 99 to 100.5% | |
| USP/EP/bioCERTIFIED |
| Clear/Colorless | |
| 0.1% | |
| 168-172°C | |
| C4H11NO3 | |
| To pass test | |
| NC(CO)(CO)CO | |
| 121.14 | |
| To pass test |