missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane 99.0+%, TCI America™

Catalog No. T251625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T251625G 25 g
T2516500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T251625G Supplier TCI America Supplier No. T251625G
Only null left

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 170°C
Color White
pH 10.0 to 10.8 (0.1mol/L aq 25°C)
Boiling Point 220°C
Quantity 25 g
Formula Weight 121.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane [for Electrophoresis]
EINECSNumber (2)-0318
RTECSNumber TY2900000
TSCA Yes