missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane Acetate 99.0+%, TCI America™

Catalog No. p-7137681
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T329425G 25 g
T3294100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T329425G Supplier TCI America Supplier No. T329425G
Only null left

Chemical Identifiers

CAS 6850-28-8
Molecular Formula C6H15NO5
Molecular Weight (g/mol) 181.188
MDL Number MFCD00038951
InChI Key PIEPQKCYPFFYMG-UHFFFAOYSA-N
Synonym tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PubChem CID 16218782
ChEBI CHEBI:66869
IUPAC Name acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES CC(=O)O.C(C(CO)(CO)N)O

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 181.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane Acetate