missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)aminomethane, 99%

Catalog No. AAA1849422
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1849422 100 g
AAA1849436 500 g
AAA184940E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1849422 Supplier Thermo Scientific Chemicals Supplier No. A1849422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis.

Solubility
Soluble in water, ethylene glycol, methanol, ethanol,dimethyl formamide, acetone, ethyl acetate, olive oil, chloroform and alkaline solution.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Density 1.353
Boiling Point 219°C to 220°C (10 mmHg)
Linear Formula (HOCH2)3CNH2
Quantity 100 g
Beilstein 741883
Sensitivity Hygroscopic
Merck Index 14,9772
Solubility Information Soluble in water,ethylene glycol,methanol,ethanol,dimethyl formamide,acetone,ethyl acetate,olive oil,chloroform and alkaline solution.
Formula Weight 121.14
Percent Purity 99%
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only