missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, 99.85%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-3779848
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327360010 1 kg Plastic Bottle
AC327360050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327360010 Supplier Thermo Scientific Chemicals Supplier No. 327360010
Only null left

CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym trometamol, tris, tromethamine, tris hydroxymethyl aminomethane, tham, trisamine, tris buffer, trizma, 2-amino-2-hydroxymethyl propane-1,3-diol, tris base
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance (0.1M in water), (0.1 M in water) at 0nm, 0.03 max. at 260nm
Melting Point >168.5°C
Color White
pH 10.0 to 11.7 (5% soln. at 25°C)
Boiling Point 219.0°C to 220.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99.85%
Packaging Plastic Bottle
Linear Formula (HOCH2)3CNH2
Quantity 1 kg
Beilstein 04, 303
Merck Index 15, 9951
Solubility Information Solubility in water: 550g/L (25°C). Other solubilities: 79.1g/L ethylene glycol- 26g/L methanol (25°C),22g/L 95% ethanol- 14g/L dmf (25°C),2g/L acetone- 0.5g/L ethyl acetate (25°C),0.05g/L chloroform- <0.05g/L ccl4 (25°C)
Formula Weight 121.14
Percent Purity 99.85%
Grade Molecular Biology
Insoluble Matter 0.0005% max
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane, For molecular biology
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-064-4

RUO – Research Use Only